CAS RN 167305-00-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Omapatrilat, MedChemExpress
MedChemExpress Omapatrilat is a dual inhibitor of the metalloproteases ACE and NEP with Ki values of 0.64 and 0.45 nM, respectively.