CAS RN 1650550-25-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Enasidenib mesylate, MedChemExpress
MedChemExpress Enasidenib mesylate is a first-in-class, oral, potent, reversible, selective inhibitor of the IDH2 mutant enzymes.