CAS RN 164579-32-2
IUPAC Nombre:
sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-2,3-dihydro-1H-1,3-benzodiazol-1-ide hydrate
Sinónimos:
sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-2,3-dihydro-1H-1,3-benzodiazol-1-ide hydrate
Peso molecular (g/mol):
425.38
Fórmula molecular:
C16H18F2N3NaO5S
InChi Key:
QHNXIKDJFYGZTL-UHFFFAOYNA-N
SMILES:
O.[Na+].COC1=CC=NC(CS(=O)C2NC3=CC(OC(F)F)=CC=C3[N-]2)=C1OC
1
–
1
de
1
resultados
1
Pantoprazole sodium hydrate, MedChemExpress
MedChemExpress Pantoprazole sodium hydrate (BY10232 sodium hydrate) is an orally active and potent proton pump inhibitor (PPI).