CAS RN 1638241-89-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ADU-S100, MedChemExpress
MedChemExpress ADU-S100 (MIW815), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity.