CAS RN 1622921-15-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Remodelin hydrobromide, MedChemExpress
MedChemExpress Remodelin, a specific inhibitor of N-acetyltransferase NAT10, can ameliorate Hutchinson-Gilford Progeria Syndrom (HGPS) cellular phenotypes. Remodelin acts in a progerin- and FTI-independent pathway, by targeting and inhibiting NAT10. NAT10 is a protein with histone acetylation activity and primarily identified to be involved in regulation of telomerase activity.