CAS RN 1598383-40-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EPZ011989, MedChemExpress
MedChemExpress EPZ011989 is a potent, selective orally bioavailable EZH2 inhibitor with Ki 3000-fold selectivity over other HMTase.