CAS RN 159752-10-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
5
de
5
resultados
1
MK 677, TRC
CAS: 159752-10-0 Sinónimo: L-163,191∣MK-0677
2
Tocris Bioscience™ MK 0677
High affinity ghrelin receptor agonist
3
Ibutamoren Mesylate, MedChemExpress
MedChemExpress Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Ibutamoren Mesylate is an orally active growth hormone (GH) secretagogue.
4
5