CAS RN 157341-41-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DMP 777, MedChemExpress
MedChemExpress DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.