CAS RN 156840-13-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ HBC 530
GFP fluorophore mimic for imaging RNA in live cells; activated by binding to Pepper aptamers; suitable for confocal and two-photon microscopy