CAS RN 150810-68-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress 5-TAMRA-SE
MedChemExpress 5-TAMRA-SE is an amine-reactive fluorescent agent, its conjugates yield bright, pH-insensitive orange-red fluorescence (approximate excitation/emission maxima ~546/579) with good photostability.
2
Tocris Bioscience™ 5-TAMRA, NHS
Fluorescent amine-reactive dye for labeling proteins, peptides and oligonucleotides