CAS RN 149155-45-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress CREBtide
MedChemExpress CREBtide, a synthetic 13 amino acid peptide, has been reported as a PKA substrate.
2
MedChemExpress CREBtide
MedChemExpress CREBtide, a synthetic 13 amino acid peptide, has been reported as a PKA substrate.
3
MedChemExpress CREBtide
MedChemExpress CREBtide, a synthetic 13 amino acid peptide, has been reported as a PKA substrate.