CAS RN 146801-07-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Promociones disponibles
Thermo Scientific™ High–Purity Fos-Choline Detergents
Thermo Scientific™ High–Purity Fos-Choline Detergents
Fos-choline detergents are a class of zwitterionic detergents commonly used in protein solubilization for various applications, including membrane protein research, protein purification, and structural biology studies.