CAS RN 1449779-49-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DGAT1-IN-1, MedChemExpress
MedChemExpress DGAT1-IN-1 is a potent DGAT1 inhibitor with IC50 of < 10 nM(cell lysate from Hep3B cells overexpressing human DGAT1).