CAS RN 1448867-42-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Siremadlin (R Enantiomer), MedChemExpress
MedChemExpress Siremadlin R Enantiomer (NVP-HDM201 R Enantiomer) is the R enantiomer of Siremadlin. Siremadlin is a potent and highly specific MDM-2/p53 inhibitor.