CAS RN 144678-63-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
6
de
6
resultados
1
Tocris Bioscience™ NSC 668036
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
2
Tocris Bioscience™ Ex 26
Potent and selective S1P1 receptor antagonist
3
Tocris Bioscience™ NSC 668036
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
4
Tocris Bioscience™ NSC 668036
Wnt signaling inhibitor; binds the PDZ domain of the Dishevelled (Dvl) protein
5
6