CAS RN 14417-88-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Melinamide, MedChemExpress
MedChemExpress Melinamide, an amide derivative of an unsaturated long-chain fatty acid, is an inhibitor of cholesterol absorption with an IC50 of 20.9 μM.