CAS RN 140147-77-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Epimedin A1, MedChemExpress
MedChemExpress Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines.