CAS RN 1334298-90-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Itacitinib, MedChemExpress
MedChemExpress Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2; Itacitinib is used in the research of myelofibrosis.