CAS RN 1233948-61-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
EL-102, MedChemExpress
MedChemExpress EL102 is a inhibitor of HIF1α , Which can inhibit tubulin polymerisation and decreased microtubule stability.