CAS RN 1227301-51-0
IUPAC Nombre:
N-[1-({7-[(4-aminobutyl)amino]heptyl}carbamoyl)-2-(4-hydroxyphenyl)ethyl]butanamide dihydrochloride
Sinónimos:
N-[1-({7-[(4-aminobutyl)amino]heptyl}carbamoyl)-2-(4-hydroxyphenyl)ethyl]butanamide dihydrochloride
Peso molecular (g/mol):
507.54
Fórmula molecular:
C24H44Cl2N4O3
InChi Key:
HWTJQQMIKVJWLH-UHFFFAOYNA-N
SMILES:
Cl.Cl.CCCC(=O)NC(CC1=CC=C(O)C=C1)C(=O)NCCCCCCCNCCCCN
1
–
1
de
1
resultados
1
Philanthotoxin 74 dihydrochloride, MedChemExpress
MedChemExpress Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively.