CAS RN 1214265-56-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
WZ-3146, MedChemExpress
MedChemExpress WZ3146 is a mutant selective EGFR inhibitor with IC50s of 2, 2, 5, 14 and 66 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750, EGFRE746_A750/T790M and EGFR, respectively.