CAS RN 1193314-23-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Cipargamin, MedChemExpress
MedChemExpress Cipargamin (NITD609) is an potent antimalarial compound, with an IC50 of appr 1 nM against P. falciparum.