CAS RN 1141777-14-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Emixustat, MedChemExpress
MedChemExpress Emixustat, a novel visual cycle modulator, is an inhibitor of the visual cycle isomerase with an IC50 value of 4.4 nM in vitro.