CAS RN 1135243-19-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
R&D Systems™ VU 0255035
A Tocris product, part of R&D Systems. Highly selective M1 antagonist