CAS RN 1129403-56-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Amcasertib, MedChemExpress
MedChemExpress Amcasertib (BBI503) is an orally active and small-molecule multi-kinase inhibitor. Amcasertib exhibits inhibitory activity against the NANOG and CD133 expression and cell viability in PC-9/GR cells. As an orally available cancer cell stemness kinase inhibitor with potential antineoplastic activity, it is currently being studied in phase I clinical trials in a number of cancers.