CAS RN 108648-07-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Purpureaside C, MedChemExpress
MedChemExpress Purpureaside C is a phenolic glycoside and has significant proinflammatory action.