CAS RN 1085703-32-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Vitamin CK3, MedChemExpress
MedChemExpress Vitamin CK3 is the combination of Vitamin C and vitamin K3 (100:1 ratio) and has been shown to inhibit tumor growth and lung metastasis.