Learn More
Invitrogen™ Novex IL-4 Swine ELISA Kit
Key cytokine in the immune network
Marca: Invitrogen™ KSC0041
Detalles adicionales : Peso : 0.50000kg
Descripción
The IL-4 Swine ELISA Kit quantifies natural and recombinant swine IL-4 in serum, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Especificaciones
Serum, Supernatant | |
397225 | |
2pg/mL | |
For research use only (RUO) | |
3 hr. |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays | |
Interleukin 4 |
For Research Use Only. Not for use in diagnostic procedures.