Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture
Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation
Marca: MP Biomedicals™ 0219989691
Detalles adicionales : Peso : 0.01000kg
Descripción
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.
Physical characteristics:
- Form: Off white to brown powder
- Purity:≥98% (w/w)
- Total Protein: ≥96%
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Certifications: ISO9001, ISO13485, cGMP, EDQM
Especificaciones
Mammalian Cell Culture | |
66 | |
6.5 to 7.5 | |
1000g | |
4°C |
Lipid-rich | |
CellMaxx™ Bovine Albumin, Low Endotoxin | |
Chromatography | |
Bovine | |
Powder |