Learn More
Thermo Scientific™ Tarjetas IR desechables, 5x10 cm, ventana de NaCl, abertura de 9,5 mm
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Marca: Thermo Scientific™ 043931.KJ
Detalles adicionales : Peso : 0.01000kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Tamaño: 5 x 10 cm
- Ventana de NaCl
- abertura de 9,5 mm
Especificaciones
Tarjetas IR desechables | |
5/paquete |