Learn More
Zoniporide Hydrochloride, TRC
Product Code. 30457306
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457306 10 mg -
30449195 100 mg 100mg
30451951 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457306 Supplier Toronto Research Chemicals Supplier No. TRCZ68000010MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 241800-97-5
Molecular Formula C17H16N6O2 • (HCl)
Molecular Weight (g/mol) 356.81
Synonym N-(Aminoiminomethyl)-5-cyclopropyl-1-(5-quinolinyl)-1H-pyrazole-4-carboxamide Hydrochloride
IUPAC Name N-carbamimidoyl-5-cyclopropyl-1-(quinolin-5-yl)-1H-pyrazole-4-carboxamide hydrochloride
SMILES O=C(NC(N)=N)C(C=N1)=C(C2CC2)N1C3=CC=CC4=NC=CC=C43.Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C17H16N6O.ClH/c18-17(19)22-16(24)12-9-21-23(15(12)10-6-7-10)14-5-1-4-13-11(14)3-2-8-20-13;/h1-5,8-10H,6-7H2,(H4,18,19,22,24);1H
Formula Weight 356.12
Chemical Name or Material Zoniporide Hydrochloride