Learn More
ZO-1 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL594-21773-100UL
Detalles adicionales : Peso : 0.00010kg
Código de producto. 17254786
  • 430.00€ / 100 microlitros
Fecha estimada de envío: 07-05-2024
para ver el stock
Descripción
Descripción
Tight junction (or zonula occludens) form the continuous intercellular barrier between epithelial and endothelial cells, which is required to separate tissue spaces and regulate selective movement of solutes across the epithelium and endothelium (PMID: 20066090). ZO-1 (also known as TJP1) is a peripheral membrane phosphoprotein located on the cytoplasmic face and is expressed in tight junctions of both epithelial and endothelial cells (PMID: 3528172). It binds the transmembrane proteins occludin and the claudins linking them to cytoskeletal actin (PMID: 17418867). ZO-1 belongs to a family of multidomain proteins known as the membrane-associated guanylate kinase homologs (MAGUKs). It is a pivotal tight junction protein and may be involved in signalling mechanisms regulating cell proliferation and differentiation (PMID: 22782886).
Especificaciones
Especificaciones
ZO-1
Polyclonal
CoraLite 594
100 μL
7082
Liquid
Immunofluorescence
1000 μg/mL