Learn More
ZNF79 Antibody (OTI3D2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NBP2-74955AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215627
  • 736.48€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
ZNF79 Monoclonal specifically detects ZNF79 in Human samples. It is validated for Western Blot.
Especificaciones
Especificaciones
ZNF79
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
ZNF79
Full length recombinant protein of human ZNF79 (NP_009066) produced in E.coli.
0.1 mL
7633.0
Store at 4°C in the dark.
Western Blot
OTI3D2
Western Blot
pT7, zinc finger protein 79, zinc finger protein 79 (pT7), ZNFpT7
Mouse
Affinity Purified
Primary
Human
IgG2b