missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF611 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17249-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May be involved in transcriptional regulation.Especificaciones
ZNF611 | |
Polyclonal | |
Unconjugated | |
ZNF611 | |
zinc finger protein 611, ZNF611 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81856 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N823 | |
ZNF611 | |
ZNF611 Fusion Protein Ag9984 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |