missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11219-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintece in the progenitor stage by promoting the cell cycle. Specifically binds to the 5′-TCAT-3′ DNA sequence. Has transcription repressor activity in vitro.Especificaciones
ZNF24 | |
Polyclonal | |
Unconjugated | |
ZNF24 | |
KOX17, RSG A, Zfp191, Zinc finger protein 191, zinc finger protein 24, Zinc finger protein KOX17, ZNF191, ZNF24, ZSCAN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360204, 59057, 7572 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17028, Q7TNK3, Q91VN1 | |
Znf24 | |
ZNF24 Fusion Protein Ag1724 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |