missing translation for 'onlineSavingsMsg'
Learn More

ZNF146 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-83826-0.1ML

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216319

  • 455.71€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ZNF146 Polyclonal specifically detects ZNF146 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

ZNF146
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
7705
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
MGC125660, MGC125661, only zinc finger protein, OZF, zinc finger protein 146, zinc finger protein OZF
The immunogen is a synthetic peptide directed towards the middle region of human ZNF146
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales