missing translation for 'onlineSavingsMsg'
Learn More

ZMYM4 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-82054UV

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213206

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones

Especificaciones

ZMYM4
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262
ZMYM4 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZMYM4. The immunogen is located within the last 50 amino acids of ZMYM4. Amino Acid Squence: EVHEELAKAKSEDSDVE
0.1 mL
Cell Biology
9202
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales