Learn More
Terc-butóxido de circonio (IV), 99,99 % (base metálica), Thermo Scientific Chemicals
Descripción
It finds it application as a reagent for the preparation of heterometallic tin(II) or lead(II) zirconium alkoxides. It is used as a catalyst in cyanation process. Zirconium tert-butoxide (ZTB) precursor is used to deposit thin films of Zirconia and other zirconium containing films by atomic layer deposition and chemical vapor deposition methods. Zirconium oxide thin films can also be grown at low temperatures, ranging from 150°C to 300°C, by the presence of moisture along with ZTB or by UV-enhanced atomic layer deposition (UV-ALD) process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Zirconium(IV) tert-butoxide |
Color | Incolora |
Densidad | 0.985 g/mL |
Punto de ebullición | 90°C (5 mmHg) |
Temperatura de inflamación | 85°C (185°F) |
Notas del porcentaje del ensayo | (base metálica) |
Fórmula lineal | Zr[OC(CH3)3]4 |
Cantidad | 10 g |
Beilstein | 3681870 |
Sensibilidad | Light and moisture sensitive |
Mostrar más |
RUO – Research Use Only