missing translation for 'onlineSavingsMsg'
Learn More

Óxido de cinc, NanoTek™, Thermo Scientific Chemicals

Catalog Number p-4782241
Click to view available options
Cantidad:
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxocinc
O=[Zn]
Este artículo no se puede devolver. Vea la política de devoluciones
1314-13-2
OZn
81.38
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
14806
CHEBI:36560
oxocinc
O=[Zn]

NanoTek™ zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systemsZinc oxide nanoparticles are used in the manufacture of rubber and cigarettes. It plays an important role in paints as a coating agent. It is used in creams and ointments, which are used to treat skin diseases. It acts as an additive in the manufacture of concrete. It is also used in ceramic industry and as an additive in food products such as breakfast cereals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Zinc Oxide
Punto de fusión 1975°C
Forma física Polvo APS de 40-100 nm,S.A. de 10 a 25 m
Cantidad 100 g
Información de solubilidad Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkali hydroxide solution.
Formula Weight (peso de la fórmula) 81.37
Densidad 5.6

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'