Learn More
ZEOprep™ Diol Functionalized
Product Code. 30049002 Shop All Zeochem Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049002 5 kg 5kg
30048965 100 g 100g
30049026 1 kg 1kg
30048949 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30049002 Supplier ZEOprep Supplier No. 302091
  • ONLINE EXCLUSIVE PRICE
    4780.00€
    4520.25€ / 5 Kg
    Ahorro 259.75€
    5% Descuento
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. Diol functionalized Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. The DIOL ligand results in a less polar surface than bare silica.
  • DIOL functionalized for low polarity surface
  • Ideal for lipid applications
  • Suitable for SFC Irregular Amorphous Silica Gel
  • High Purity
  • European Manufacture
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • High Chemical and Mechanical Stability
  • Narrow Particles Size Distribution
  • 20 to 500 μm particles size range
  • Available in 60 A or 90 A pore size
  • High loadability
  • Selection of pack sizes available
  • Suitable for gravity, flash, prep and process scale
Specifications
Particle Size 40 to 63 μm
Pore Size 60 Å
Technique Chromatography
Quantity 5 kg
Product Type Silica Gel
Material Silica Gel
Phase DIOL Functionalized
Particle Shape Irregular