Learn More
Thermo Scientific™ Zeiss 40X Objective, LD A-Plan (0.5 NA)
Product Code. 17226583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity Unit Size
17226583 1 each Each
1 options
This item is not returnable. View return policy
Product Code. 17226583 Supplier Thermo Scientific™ Supplier No. N010097
 Restricted Item
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Expand the functionality of your Thermo Scientific™ high-content instrument
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
TRUSTED_SUSTAINABILITY
Specifications
Type Objective Lens
Product Line Zeiss
Description Zeiss 40X Objective, LD A-Plan (0.5 NA)
For Use With (Equipment) ArrayScan HCA Readers
Lens Type Dry
Magnification 40x (0.5NA)
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.