missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25985-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Especificaciones
ZDHHC7 | |
Polyclonal | |
Unconjugated | |
ZDHHC7 | |
DHHC 7, Palmitoyltransferase ZDHHC7, ZDHHC7, Zinc finger protein 370, ZNF370 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55625 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NXF8 | |
ZDHHC7 | |
ZDHHC7 Fusion Protein Ag23185 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |