missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H7B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25624-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.Especificaciones
ZC3H7B | |
Polyclonal | |
Unconjugated | |
ZC3H7B | |
KIAA1031, RoXaN, ZC3H7B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20286, 23264 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UGR2 | |
Zc3h7b | |
ZC3H7B Fusion Protein Ag22555 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |