missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H12A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25009-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.Especificaciones
ZC3H12A | |
Polyclonal | |
Unconjugated | |
ZC3H12A | |
MCP induced protein 1, MCPIP, MCPIP1, Ribonuclease ZC3H12A, RP3 423B22.1, ZC3H12A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230738, 80149 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5D1E7, Q5D1E8 | |
ZC3H12A | |
ZC3H12A Fusion Protein Ag13877 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |