Learn More
(Z)-rac-Luliconazole, TRC
Product Code. 30482131
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30482131 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30482131 Supplier Toronto Research Chemicals Supplier No. TRCL473780100MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (Z)-rac-Luliconazole
CAS 101529-76-4
Molecular Formula C14H9Cl2N3S2
Quantity 100 mg
Synonym (Z)-alpha-[4-(2,4-Dichlorophenyl)-1,3-dithiolan-2-ylidene]-1H-imidazole-1-acetonitrile
InChI Formula InChI=1S/C14H9Cl2N3S2/c15-9-1-2-10(11(16)5-9)13-7-20-14(21-13)12(6-17)19-4-3-18-8-19/h1-5,8,13H,7H2/b14-12-
SMILES Clc1ccc(C2CS\C(=C(/C#N)\n3ccnc3)\S2)c(Cl)c1
IUPAC Name (2Z)-2-[4-(2,4-dichlorophenyl)-1,3-dithiolan-2-ylidene]-2-imidazol-1-ylacetonitrile
Molecular Weight (g/mol) 354.28
Formula Weight 352.9615