missing translation for 'onlineSavingsMsg'
Learn More

2-Metoxietóxido de itrio(III), 5 % p/v en 2-metoxietanol, Thermo Scientific™

Catalog Number p-7051695
Click to view available options
Cantidad:
25 mL
100 mL
Este artículo no se puede devolver. Vea la política de devoluciones
115668-57-0
C9H21O6Y
314.17
MFCD00156527,MFCD00156527
IVXUPDRNSUCVNX-UHFFFAOYSA-N
1-methoxyethanolate,yttrium 3+, 2-methoxyethanolate;yttrium 3+
14452922
yttrium(3+) tris(2-methoxyethan-1-olate)
[Y+3].COCC[O-].COCC[O-].COCC[O-]
Este artículo no se puede devolver. Vea la política de devoluciones
115668-57-0
C9H21O6Y
314.17
MFCD00156527,MFCD00156527
IVXUPDRNSUCVNX-UHFFFAOYSA-N
1-methoxyethanolate,yttrium 3+, 2-methoxyethanolate;yttrium 3+
14452922
yttrium(3+) tris(2-methoxyethan-1-olate)
[Y+3].COCC[O-].COCC[O-].COCC[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Yttrium(III) 2-methoxyethoxide
Fórmula lineal Y(OCH2CH2OCH3)3
Cantidad 25 mL
Número UN UN1188
Sensibilidad Moisture sensitive
Formula Weight (peso de la fórmula) 314.165
Concentration or Composition (by Analyte or Components) 5 % p/v en 2-metoxietanol
Forma física Líquida
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'