Learn More
YSI™ ODO Sensor Cap for ProODO Optical Dissolved Oxygen Meter
Product Code. 11968736
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
11968736 Each
1 options
This item is not returnable. View return policy
Product Code. 11968736 Supplier YSI™ Supplier No. 626320
 Restricted Item
Fecha estimada de envoi: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Sensor cap for use with YSI ProODO Handheld Optical Dissolved Oxygen Meter (Cat. No. 14-660-204).
Replacement accessory for PRO ODO screw-on sensors. Includes: screw-on sensor cap, calibration code sequence.
TRUSTED_SUSTAINABILITY
Specifications
Includes Screw-on sensor cap and calibration code sequence