Learn More
Invitrogen™ Human YES1, GST Tag Recombinant Protein
YES1 is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
Marca: Invitrogen™ A15559
Detalles adicionales : Peso : 0.10000kg
Descripción
- Active—this protein can be used in kinase activity assays, such as Z’-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA).
- Purified—purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA).
- GST-tagged—provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody.
Especificaciones
NP_005424 | |
88.3 kDa | |
YES1 | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Kinases & Inhibitors | |
Frozen |
Kinase Assay | |
1mg | |
SRC Kinase Family | |
Recombinant | |
50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol. For maximum recovery please spin prior to use. Aliquoting the 5 μg, 10 μg, and 20 μg sizes of kinase is not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Never store a kinase diluted. If properly stored at -80°C, this product is guaranteed for 6 months from date of purchase. |
|
Baculovirus | |
Full Length | |
Drug Development |