Learn More
Checkers Yellow Jacket™ AMS™ 5 Channel Center
Product Code. 17772428
Change view
Click to view available options
Color:
Black, Yellow
Unit Size:
Each
Product Code. Color Unit Size
17772428 Black, Yellow Each
1 options
This item is not returnable. View return policy
Product Code. 17772428 Supplier Checkers Supplier No. PYJS0004YB
  • ONLINE EXCLUSIVE PRICE
    323.17€
    262.23€ / paquete de 1
    Ahorro 60.94€
    18% Descuento
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Yellow Jacket™ 5-channel AMS center section cable protector is a patented technology that utilizes separate cable protector sections and ramps to create a system that is fully expandable to hold an unlimited number of cables.
  • Yellow Jacket™ 5-channel AMS™ center section cable protector is a patented technology that utilizes separate cable protector sections and ramps to create a system that is fully expandable to hold an unlimited number of cables
  • By detaching the ramps, you can also quickly add cable runs and ADA compatible side ramps to existing systems
  • The Yellow Jacket™ AMS™ readily handles heavy truck and bus traffic while providing easy access for pedestrians, carts and wheelchairs
  • Hinged lids make for easy loading of cables and hoses
Specifications
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 25 cm
Color Black, Yellow
Material Polyethylene
Type 5 Channel Center