Learn More
YEATS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24717-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The YEATS domain-containing protein 2 (YEATS2) is a nuclear protein that is phosphorylated upon DNA damage by ATM or ATR and contains 1 YEATS domain. The gene encoding YEATS2 resides on chromosome 3, which contains about 214 million bases encoding over 1, 100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B.Especificaciones
YEATS2 | |
Polyclonal | |
Unconjugated | |
YEATS2 | |
KIAA1197, YEATS domain containing 2, YEATS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55689 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ULM3 | |
YEATS2 | |
YEATS2 Fusion Protein Ag20134 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |